missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Keap1 Antibody (OTI1B4), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71496AF700
This item is not returnable.
View return policy
Description
Keap1 Monoclonal specifically detects Keap1 in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.
Specifications
| Keap1 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| KEAP1 | |
| Full length recombinant protein of human Keap1 (NP_987096) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| OTI1B4 | |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| INRF2, INrf2MGC9454, kelch-like ECH-associated protein 1, Kelch-like protein 19, KIAA0132MGC4407, KLHL19Cytosolic inhibitor of Nrf2, MGC10630, MGC1114, MGC20887 | |
| Mouse | |
| Affinity Purified | |
| Stem Cell Markers | |
| 9817.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction