Learn More
DWK Life Sciences™ KECK™ Tubing Clamp, PBT, KT
Product Code. 15844688 Shop All DWK Life Sciences Products
Click to view available options
Size:
Fits tube OD 10 mm
Fits tube OD 14 mm
Fits tube OD 4.5 mm
Fits tube OD 6 mm
Unit Size:
Each
Pack of 100
Product Code. 15844688
Brand: DWK Life Sciences™ 286338204
For infinitely variable regulation of flow rates.
With just one finger, flow rates in hoses can be infinitely adjusted with KECK™ hose clamps. The clamps are available in different colours for the different hose diameters.
  • Continuous regulation of flow rates
  • Handling with one hand possible
  • Color coding of the clamps according to the different hose diameters
TRUSTED_SUSTAINABILITY
Specifications
Quantity 100
Material PBT
For Use With (Equipment) Infinitely Variable Regulation of Flow Rates
For Use With (Application) infinitely variable regulation of flow rates
Size Fits tube OD 6 mm
Color Yellow