missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KEL Mouse anti-Human, Clone: OTI5E6, liquid, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811547
This item is not returnable.
View return policy
Description
This gene encodes a type II transmembrane glycoprotein that is the highly polymorphic Kell blood group antigen. The Kell glycoprotein links via a single disulfide bond to the XK membrane protein that carries the Kx antigen. The encoded protein contains sequence and structural similarity to members of the neprilysin (M13) family of zinc endopeptidases.Specifications
| KEL | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P23276 | |
| KEL | |
| Human recombit protein fragment corresponding to amino acids 166-420 of human KEL produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI5E6 | |
| Unconjugated | |
| KEL | |
| CD238, ECE3 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 3792 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction