missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kelch Domain Containing 7A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 391.65 - € 556.50
Specifications
| Antigen | Kelch Domain Containing 7A |
|---|---|
| Dilution | Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1-4 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18450742
|
Novus Biologicals
NBP2-30941-25ul |
25 μL |
€ 415.00 € 391.65 / 25µL Save € 23.35 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18130205
|
Novus Biologicals
NBP2-30941 |
0.1 mL |
€ 589.00 € 556.50 / 0.10mL Save € 32.50 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Kelch Domain Containing 7A Polyclonal specifically detects Kelch Domain Containing 7A in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| Kelch Domain Containing 7A | |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| Kelch Domain-Containing Protein 7A, KLHDC7A, RP11-422P22.2 | |
| KLHDC7A | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1-4 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| Q5VTJ3 | |
| 127707 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: CATYRTPYPDAFQCAVVDNLIYCVGRRSTLCFLADSVSPRFVPKELRSFPAPQGTLLPTVLTLPTPDLPQTR | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit