missing translation for 'onlineSavingsMsg'
Learn More

Kelch-Like Family Member 21 Antibody (2H4), Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-37659-0.025ml

 View more versions of this product

Product Code. 18655215

  • € 203.00 / 0.03mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Kelch-Like Family Member 21 Monoclonal specifically detects Kelch-Like Family Member 21 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Kelch-Like Family Member 21
Monoclonal
Unconjugated
Ascites with 0.03% Sodium Azide
KLHL21
Purified recombinant fragment of human KLHL21 expressed in E. Coli.
Unpurified
RUO
9903
Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles.
IgG1
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
2H4
Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200-1:1000
kelch-like 21 (Drosophila), Kelch-Like Protein 21, KIAA0469, KLHL21
Mouse
66 kDa
0.025 mL
Primary
Human
Ascites
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only