Learn More
Invitrogen™ KELL Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5139866
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: PRPCETSVCL DLRDHYLASG NTSVAPCTDF FSFACGRAKE TNNSFQELAT KNKNRLRRIL EVQNSWHPGS GEEKAFQFYN SCMDTLAIEA AGTGPLRQVI EELGGWRISG KWT Highest antigen sequence indentity to the following orthologs: Rat - 70%, Mouse - 73%.
This gene encodes a type II transmembrane glycoprotein that is the highly polymorphic Kell blood group antigen. The Kell glycoprotein links via a single disulfide bond to the XK membrane protein that carries the Kx antigen. The encoded protein contains sequence and structural similarity to members of the neprilysin (M13) family of zinc endopeptidases.
Specifications
| KELL | |
| Polyclonal | |
| Unconjugated | |
| KEL | |
| CD238; ECE3; Kel; Kel blood group protein; Kell blood group; kell blood group antigen; kell blood group glycoprotein; Kell blood group glycoprotein homolog; Kell blood group, metallo-endopeptidase; Kell protein; RGD1565470 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 3792 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P23276 | |
| KEL | |
| Recombinant protein corresponding to Human KEL. Recombinant protein control fragment (Product #RP-109060). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.