missing translation for 'onlineSavingsMsg'
Learn More

Ketoconazole, MedChemExpress

Product Code. 16458096
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16458096 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16458096 Supplier Medchem Express Supplier No. HYB0105
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Ketoconazole (R-41400) is an imidazole anti-fungal agent, a CYP3A4 and CYP24A1 inhibitor.

Ketoconazole (R-41400) is an imidazole anti-fungal agent, a CYP3A4 and CYP24A1 inhibitor.

Specifications

Chemical Name or Material Ketoconazole
Physical Form Solid
CAS 65277-42-1
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C26H28Cl2N4O4
For Use With (Application) COVID-19-immunoregulation
Synonym Ketoconazol R 41400
Solubility Information DMSO : 25 mg/mL (47.04 mM; Need ultrasonic)
SMILES CC(N1CCN(C2=CC=C(OC[C@@H]3O[C@@](CN4C=CN=C4)(C5=CC=C(Cl)C=C5Cl)OC3)C=C2)CC1)=O
Molecular Weight (g/mol) 531.43
Formula Weight 531.43
Percent Purity 99.47%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.