Learn More
Ketoconazole N-Oxide, Mikromol™
Description
Ketoconazole N-Oxide
Specifications
Specifications
| Chemical Name or Material | Ketoconazole N-Oxide |
| CAS | 254912-65-7 |
| Analyte or Component Names | Ketoconazole N-Oxide |
| Shipping Condition | Room Temperature |
| Molecular Formula | C26 H28 Cl2 N4 O5 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Ethanone, 1-[4-[4-[[(2R,4S)-2-(2,4-dichlorophenyl)-2-(1H-imidazol-1-ylmethyl)-1,3-dioxolan-4-yl]methoxy]phenyl]-4-oxido-1-piperazinyl]-, rel-, Piperazine, 1-acetyl-4-[4-[[(2R,4S)-2-(2,4-dichlorophenyl)-2-(1H-imidazol-1-ylmethyl)-1,3-dioxolan-4-yl]methoxy]phenyl]-, 4-oxide, rel- (9CI), R 053165 |
| InChI Formula | InChI=1S/C26H28Cl2N4O5/c1-19(33)31-10-12-32(34,13-11-31)21-3-5-22(6-4-21)35-15-23-16-36-26(37-23,17-30-9-8-29-18-30)24-7-2-20(27)14-25(24)28/h2-9,14,18,23H,10-13,15-17H2,1H3/t23-,26-/m0/s1 |
| SMILES | CC(=O)N1CC[N+]([O-])(CC1)c2ccc(OC[C@H]3CO[C@@](Cn4ccnc4)(O3)c5ccc(Cl)cc5Cl)cc2 |
| IUPAC Name | 1-[4-[4-[[(2R,4S)-2-(2,4-dichlorophenyl)-2-(imidazol-1-ylmethyl)-1,3-dioxolan-4-yl]methoxy]phenyl]-4-oxidopiperazin-4-ium-1-yl]ethanone |
| Show More |