Learn More
DiscoverX α-Ketoglutaric acid sodium salt
Product Code. 17382529
Encompass
Click to view available options
:
10 mg
Product Code. 17382529
Brand: DiscoverX 921212
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
α-Ketoglutaric acid is a naturally occuring ketone derivitive of glutaric acid and intermediate of the krebs cycle. It serves as a nitrogen transporter, antioxidant, anabolic, wound healer, and may have anti-aging properties.
α-Ketoglutaric acid is a naturally occuring ketone derivitive of glutaric acid and intermediate of the krebs cycle. It serves as a nitrogen transporter, antioxidant, anabolic, wound healer, and may have anti-aging properties. Used to treat kidney disease, GI disorders, bacterial infections, liver problems, cataracts, and yeast infections. Commonly sold as a supplement to help combat ammonia toxicity.
Specifications
Quantity 10 mg