missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ketohexokinase Antibody (OTI1D8), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71069AF350
This item is not returnable.
View return policy
Description
Ketohexokinase Monoclonal specifically detects Ketohexokinase in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| Ketohexokinase | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| KHK | |
| Full length recombinant protein of human Ketohexokinase (NP_000212) produced in HEK293T cells. | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI1D8 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| EC 2.7.1.3, Hepatic fructokinase, ketohexokinase, ketohexokinase (fructokinase) | |
| Mouse | |
| 0.1 mL | |
| 3795 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction