missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ketohexokinase Antibody (OTI1D8), Allophycocyanin/Cy7, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71069APCCY7
This item is not returnable.
View return policy
Description
Ketohexokinase Monoclonal specifically detects Ketohexokinase in Human samples. It is validated for Flow Cytometry.
Specifications
| Ketohexokinase | |
| Monoclonal | |
| APC-Cyanine7 | |
| PBS with 0.05% Sodium Azide | |
| KHK | |
| Full length recombinant protein of human Ketohexokinase (NP_000212) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Flow Cytometry | |
| OTI1D8 | |
| Flow Cytometry | |
| EC 2.7.1.3, Hepatic fructokinase, ketohexokinase, ketohexokinase (fructokinase) | |
| Mouse | |
| Affinity Purified | |
| Lipid and Metabolism | |
| 3795.0 | |
| Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction