missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ketohexokinase Antibody (OTI1D8), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71069CL1
This item is not returnable.
View return policy
Description
Ketohexokinase Monoclonal antibody specifically detects Ketohexokinase in Human samples. It is validated for Flow Cytometry,Immunohistochemistry,Western Blot,CyTOF,Immunocytochemistry/Immunofluorescence
Specifications
| Ketohexokinase | |
| Monoclonal | |
| CoraFluor 1 | |
| EC 2.7.1.3, Hepatic fructokinase, ketohexokinase, ketohexokinase (fructokinase) | |
| Full length human recombinant protein of human KHK (NP_000212) produced in HEK293T cell. | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 3795 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry, Western Blot, CyTOF, Immunocytochemistry/Immunofluorescence | |
| OTI1D8 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction