Learn More
Ketoprofen EP Impurity A (Ketoprofen USP Related Compound D), Mikromol™
Product Code. 17909989
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17909989 25 mg 25mg
18090160 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17909989 Supplier Mikromol Supplier No. MM0001.010025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Ketoprofen EP Impurity A (Ketoprofen USP Related Compound D)
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 3-Acetylbenzophenone
CAS 66067-44-5
Analyte or Component Names 3-Acetylbenzophenone
Shipping Condition Room Temperature
Molecular Formula C15 H12 O2
Certifications/Compliance ISO 17025
Synonym 1-(3-Benzoylphenyl)ethanone (3-Acetylbenzophenone), Ketoprofen Imp. A (EP), Ketoprofen USP Related Compound D, Ketoprofen USP RC D, 1-(3-Benzoylphenyl)ethanone, 3-Acetylbenzophenone, Ketoprofen Related Compound D, Ketoprofen Impurity A
InChI Formula InChI=1S/C15H12O2/c1-11(16)13-8-5-9-14(10-13)15(17)12-6-3-2-4-7-12/h2-10H,1H3
SMILES CC(=O)c1cccc(c1)C(=O)c2ccccc2
IUPAC Name 1-(3-benzoylphenyl)ethanone
Molecular Weight (g/mol) 224.25
Quantity 25 mg
Formula Weight 224.0837 g/mol