Learn More
Tocris Bioscience™ KG 5
GREENER_CHOICE
PDGFRβ, B-Raf, c-Raf, FLT3 and KIT inhibitor
Brand:  Tocris Bioscience™ 5260/50
Encompass
Product Code. 15766626
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
KG 5 is a PDGFRβ and B-Raf allosteric inhibitor. Also inhibits c-Raf, FLT3 and KIT. Disrupts blood vessel formation in zebrafish embryos and inhibits angiogenesis in a mouse angiogenesis model. Inhibits growth of renal and pancreatic tumors in mice. Also arrests cells in prometaphase. Orally available.
Specifications
Specifications
2-(Methylthio)-6-[4-[5-[[3-(trifluoromethyl)phenyl]amino]-1H-1,2,4-triazol-3-yl]phenoxy]-4-pyrimidinamine
877874-85-6