missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ KG 5
GREENER_CHOICE

Product Code. 18780789
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18780789 50 mg 50mg
18780518 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18780789 Supplier Tocris Bioscience™ Supplier No. 5260/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

PDGFRβ, B-Raf, c-Raf, FLT3 and KIT inhibitor

KG 5 is a PDGFRβ and B-Raf allosteric inhibitor. Also inhibits c-Raf, FLT3 and KIT. Disrupts blood vessel formation in zebrafish embryos and inhibits angiogenesis in a mouse angiogenesis model. Inhibits growth of renal and pancreatic tumors in mice. Also arrests cells in prometaphase. Orally available.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-(Methylthio)-6-[4-[5-[[3-(trifluoromethyl)phenyl]amino]-1H-1,2,4-triazol-3-yl]phenoxy]-4-pyrimidinamine
CAS 877874-85-6
Quantity 50 mg
Target Raf Kinase Inhibitors
Molecular Formula C20H16F3N7OS
Formula Weight 459.45
Purity >98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.