missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress KGF/FGF-7 Protein, Human (CHO)

Product Code. 16429246
Encompass_Preferred
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
This item is not returnable. View return policy

Product Code. 16429246

Brand: MedChemExpress HYP7047A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress KGF/FGF-7 Protein, Human (CHO) is a polypeptide mitogen that belongs to the family of fibroblast growth factors, binds only to a splice variant of FGFR2 (FGFR2 IIIb) and is a highly specific paracrine growth factor for epithelial cells. KGF/FGF-7 Protein and its receptor are important for normal wound healing.

Specifications

Content And Storage Stored at -20°C. After reconstitution, it is stable at 4°C for 1 week or -20°C for longer. It is recommended to freeze aliquots at -20°C or -80°C for extended storage.
For Use With (Application) Neuroscience-Neuromodulation
Quantity 100 μg
Shipping Condition Room temperature
Molecular Weight (g/mol) 16-18 kDa
Synonym rHuKGF-1/FGF-7∣ Keratinocyte Growth Factor∣ Fibroblast Growth Factor-7∣ HBGF-7
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.