Learn More
Invitrogen™ KGF Recombinant Rabbit Monoclonal Antibody (23GB3840), Invitrogen™
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA552580
 View more versions of this product
Product Code. 30089798
  • € 468.00 / 100µL
Description
Plays an important role in the regulation of embryonic development, cell proliferation and cell differentiation. Required for normal branching morphogenesis. Growth factor active on keratinocytes. Possible major paracrine effector of normal epithelial cell proliferation.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
KGF
Recombinant Monoclonal
1.5 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P21781, P36363, Q02195
FGF7
A synthetic peptide of human KGF (30-75AA).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Flow Cytometry, Western Blot, Immunocytochemistry
23GB3840
Unconjugated
FGF7
Fgf7; FGF-7; Fibroblast growth factor 7; HBGF-7; Heparin-binding growth factor 7; keratinocyte growth factor; Kgf
Rabbit
Affinity chromatography
RUO
14178, 2252, 29348
-20°C