missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KHDC1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-06228-50ul
This item is not returnable.
View return policy
Description
KHDC1 Polyclonal antibody specifically detects KHDC1 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
Specifications
| KHDC1 | |
| Polyclonal | |
| Immunocytochemistry/ Immunofluorescence 1:100-1:500 | |
| bA257K9.4, C6orf148, chromosome 6 open reading frame 148, KH homology domain containing 1, KH homology domain-containing protein 1, MGC10818, NDG1 | |
| Produced in rabbits immunized with E. coli-derived Human KHDC1 fragment. | |
| 50 μg | |
| Apoptosis | |
| 80759 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunofluorescence | |
| Unconjugated | |
| PBS, pH7.0 | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction