missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KHDC3L Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
€ 145.00 - € 410.00
Specifications
| Antigen | KHDC3L |
|---|---|
| Concentration | 0.47 mg/mL |
| Applications | Immunofluorescence, Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16803515
|
Proteintech
24989-1-AP-20UL |
20 μL |
€ 145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16893505
|
Proteintech
24989-1-AP-150UL |
150 μL |
€ 410.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The protein encoded by this gene belongs to the KHDC1 family, members of which contain an atypical KH domain that may not bind RNA like canonical KH domains. This gene is specifically expressed in the oocytes, and recent studies suggest that it may function as a regulator of genomic imprinting in the oocyte. Mutations in this gene are associated with recurrent biparental complete hydatidiform mole.Specifications
| KHDC3L | |
| Immunofluorescence, Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q587J8 | |
| 154288 | |
| KHDC3L Fusion Protein Ag21790 | |
| Primary | |
| -20°C | |
| KHDC3L |
| 0.47 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| C6orf221, ECAT1 | |
| KHDC3L | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title