missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KHK Polyclonal Antibody

Description
Recommended positive controls: HeLa, HepG2, mouse Liver. Predicted reactivity: Mouse (87%), Rat (86%), Dog (85%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
KHK encodes the gene ketohexokinase that catalyzes conversion of fructose to fructose-1-phosphate. The splice variant presented encodes the highly active form found in liver. renal cortex. and small intestine. while the alternate variant encodes the lowe.
Specifications
Specifications
| Antigen | KHK |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.4 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7 |
| Gene | KHK |
| Gene Accession No. | P50053, P97328, Q02974 |
| Gene Alias | fructokinase; hepatic fructokinase; KETHPRO; ketohexokinase; ketohexokinase (fructokinase); ketohexokinase isoform a; Khk; testicular tissue protein Li 102 |
| Gene Symbols | KHK |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?