missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KHSRP Antibody, FITC, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-18911F
This item is not returnable.
View return policy
Description
KHSRP Polyclonal antibody specifically detects KHSRP in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| KHSRP | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| FBP2far upstream element-binding protein 2, FUBP2p75, FUSE binding protein 2, FUSE-binding protein 2, KH type-splicing regulatory protein, KH-type splicing regulatory protein, KSRPMGC99676 | |
| The immunogen recognized by this antibody maps to a region between residue 660 and 710 of human KH-type splicing regulatory protein using the numbering given in entry Q92945.3 (GeneID 8570). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| FITC | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 8570 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction