missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Ki 20227
GREENER_CHOICE

Product Code. 18004552
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18004552

Brand: Tocris Bioscience™ 4481/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

c-Fms inhibitor; suppresses osteolysis and osteoclast accumulation

Inhibitor of c-Fms tyrosine kinase (M-CSFR, CSF1R) (IC50 values are 2, 12, 217 and 451 nM for c-Fms, VEGFR-2, PDGFRβ and c-Kit respectively). Does not inhibit Flt3, EGFR or c-Src. Suppresses osteoclast differentiation and osteolysis in a rat bone metastasis model.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 623142-96-1
Molecular Formula C24H24N4O5S

Specifications

Quantity 50 mg
Formula Weight 480.54
Purity >97%
Molecular Weight (g/mol) 480.539
Chemical Name or Material Ki 20227
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.