missing translation for 'onlineSavingsMsg'
Learn More

KI696, MedChemExpress

Product Code. 16460729
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
Unit Size:
10mg
1mg
25mg
5mg
This item is not returnable. View return policy

Product Code. 16460729

Brand: Medchem Express HY101140

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress KI696 is a high affinity probe that disrupts the Keap1/NRF2 interaction. KI696 is a potent and selective inhibitor of the KEAP1/NRF2 interaction.

KI696 is a high affinity probe that disrupts the Keap1/NRF2 interaction. KI696 is a potent and selective inhibitor of the KEAP1/NRF2 interaction.

Specifications

Chemical Name or Material KI696
CAS 1799974-70-1
Color Off-White
Purity Grade Notes Research
Molecular Formula C28H30N4O6S
Quantity 1 mg
Solubility Information DMSO : 50 mg/mL (90.81 mM; Need ultrasonic)
SMILES COC1=C(N2C)C(N=N2)=CC([C@H](C3=CC(CN(C[C@H]4C)S(=O)(C5=CC=CC=C5O4)=O)=C(C)C=C3)CC(O)=O)=C1
Molecular Weight (g/mol) 550.63
Formula Weight 550.63
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.