missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIAA0040 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 183.00 - € 322.00
Specifications
| Antigen | KIAA0040 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18659066
|
Novus Biologicals
NBP3-12664-100ul |
100 μg |
€ 322.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18697198
|
Novus Biologicals
NBP3-12664-50ul |
50 μg |
€ 183.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
KIAA0040 Polyclonal antibody specifically detects KIAA0040 in Human samples. It is validated for Western BlotSpecifications
| KIAA0040 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| FLJ75822, hypothetical protein LOC9674, KIAA0040, MGC133301 | |
| Produced in rabbits immunized with E. coli-derived Human KIAA0040 fragment | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS, pH7.0 | |
| 9674 | |
| IgG | |
| Antigen and protein A Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title