Learn More
KIAA0317 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 25623-1-AP-20UL
 View more versions of this product
Product Code. 18009385
  • € 145.00 / 20µL
Description
The KIAA0317 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human KIAA0317. This antibody recognizes human, mouse antigen. The KIAA0317 antibody has been validated for the following applications: IHC, ELISA analysis.
Specifications
Specifications
KIAA0317
400 μg/mL
Rabbit
Primary
Human, Mouse
150 kDa
IgG
Immunohistochemistry, ELISA
Apoptosis-resistant E3 ubiquitin protein ligase 1, Apoptosis-resistant HECT-type E3 ubiquitin transferase 1, AREL1, KIAA0317, Protein KIAA0317
20 μL
9870
Store at -20°C. Stable for one year after shipment.