Learn More
KIAA0317 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25623-1-AP-20UL
Description
The KIAA0317 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human KIAA0317. This antibody recognizes human, mouse antigen. The KIAA0317 antibody has been validated for the following applications: IHC, ELISA analysis.
Specifications
| KIAA0317 | |
| 400 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse | |
| 150 kDa | |
| IgG |
| Immunohistochemistry, ELISA | |
| Apoptosis-resistant E3 ubiquitin protein ligase 1, Apoptosis-resistant HECT-type E3 ubiquitin transferase 1, AREL1, KIAA0317, Protein KIAA0317 | |
| 20 μL | |
| 9870 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |