missing translation for 'onlineSavingsMsg'
Learn More

KIAA0467 Mouse anti-Human, Purified MaxPab™ Polyclonal , Abnova™

Product Code. 16168488
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16168488 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16168488 Supplier Abnova Supplier No. H00023334B01P.50ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse polyclonal antibody raised against a full-length human KIAA0467 protein.

Sequence: MLFDPAAWLHGPPETSGPPDGQRRHRPESGSGSREAPTSCESLDVSPPGAREEPWLKELSLAFLQQYVQYLQSIGFVLVPLRPPSPARSTSRPRAMAILGTEGRGSFSCPKTKTDGSPKSTSSPVTTYHLQRALPGGIILMELAFQGCYFCVKQFALECSRIPMGQAVNSQLSMLFTEECDKVRDLMHVHSFSYDFHLRLVHQHVLGAHLVLRHGYHLTTFLRHFLAHHPDGPHFGRNHIYQGTLELPTPLIAAHQLYNYVADHASSYHMKPLRMARPGGPEHNEYALVSAWHSSGSYLDSEGLRHQDDFDVSLLVCHCAAPFEEQGEAERHVLRLQFFVVLTSQRELFPRLTADMRRFRKPPRLPPEPEAPGSSAGSPGEASGLILAPGPAPLFPPLAAEVGMARARLAQLVRLAGGHCRRDTLWKRLFLLEPPGPDRLRLGGRLALAELEELLEAVHAKSIGDIDPQLDCFLSMTVSWYQSLIKVLLSRFPQSCRHFQSPDLGTQYLVVLNQKFTDCFVLVFLDSHLGKTSLTVVFREPFPVQPQDSESPPAQLVSTYHHLESVINTACFTLWTRLL

Specifications

Antigen KIAA0467
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human KIAA0467 protein.
Formulation PBS with no preservative; pH 7.4
Gene SZT2
Gene Accession No. BC052802
Gene Alias FLJ10387/FLJ23425/RP11-506B15.3
Gene Symbols KIAA0467
Host Species Mouse
Immunogen KIAA0467 (AAH52802,1 a.a. ∽ 579 a.a) full-length human protein.
Purification Method Protein A/G purification
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23334
Test Specificity Antibody reactive against mammalian transfected lysate.
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.