Learn More
NBN Monoclonal Antibody (OTI3C12), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA801448
Encompass_Preferred
 View more versions of this product
Product Code. 17599812
  • POA / 100µL
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Description
Description
The p95 gene (identical to NBS1 and nibrin) is a member of the hMre11/hRad50 double-strand break complex. This protein complex has been implicated in Nijmegen breakage syndrome, an autosomal recessive disorder marked by increased cancer incidence, cell cycle checkpoint deficits, and ionizing radiation sensitivity, thus revealing a direct molecular link between double-strand break repair and cell cycle checkpoint functions.
Specifications
Specifications
NBN
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
O60934
NBN
Human recombit protein fragment corresponding to amino acids 183-460 of human NBN produced in E.coli.
100 μL
Primary
Human
Antibody
IgG2a
Immunohistochemistry (Paraffin), Western Blot
OTI3C12
Unconjugated
NBN
AT-V1, AT-V2, ATV, NBS, NBS1, P95
Mouse
Affinity Chromatography
RUO
4683
-20° C, Avoid Freeze/Thaw Cycles