missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Complement C7 Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-00202H
This item is not returnable.
View return policy
Description
Complement C7 Polyclonal specifically detects Complement C7 in Human samples. It is validated for ELISA.
Specifications
| Complement C7 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 730 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| HRP | |
| complement component 7, complement component C7 | |
| Produced in rabbits immunized with purified, recombinant Complement C7 (P10643; Met1-Gln843). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction