Learn More
KIAA1109 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 26349-1-AP-20UL
 View more versions of this product
Product Code. 18049475
  • € 145.00 / 20µL
Description
The KIAA1109 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human KIAA1109. This antibody recognizes human, mouse antigen. The KIAA1109 antibody has been validated for the following applications: IHC, ELISA analysis.
Specifications
Specifications
KIAA1109
700 μg/mL
Rabbit
Primary
Human, Mouse
150 kDa
IgG
Immunohistochemistry, ELISA
FSA, KIAA1109, KIAA1371
20 μL
84162
Store at -20°C. Stable for one year after shipment.