missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KIAA1324L Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA567405
This item is not returnable.
View return policy
Description
Immunogen sequence: YFHFFNISLCG HEGKKMALCT NNITDFTVKE IVAGSDDYTN LVGAFVCQST IIPSESKGFR AALSSQSIIL ADTFIGVTVE TT Highest antigen sequence identity to the following orthologs - mouse 90%, rat 89%.
KIAA1324L is a protein coding gene.
Specifications
| KIAA1324L | |
| Polyclonal | |
| Unconjugated | |
| KIAA1324L | |
| 9330182L06Rik; EIG121L; EIG121-like; Estrogen-induced gene 121-like protein; h EIG121-like; hEIG121L; KIAA1324 like; KIAA1324L; KIAA1324-like; RIKEN cDNA 9330182L06 gene; UPF0577 protein KIAA1324-like; UPF0577 protein KIAA1324-like homolog | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 222223 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| A8MWY0 | |
| KIAA1324L | |
| Recombinant Human KIAA1324L. Recombinant protein control fragment (Product #RP-108070). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction