Learn More
Invitrogen™ KIAA1429 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5144071
Description
Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: human PC-3 whole cell, human SK-OV-3 whole cell, human K562 whole cell, human HEL whole cell. IHC: human bladder epithelial carcinoma tissue, human breast cancer tissue. ICC/IF: U20S cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
KIAA1429 is required for N6-methyladenosine (m6A) methylation of mRNAs, a modification that plays a role in the efficiency of mRNA splicing and processing. It is also Involved in mRNA splicing regulation, probably via its function in m6A methylation. Diseases associated with KIAA1429 include Retinitis Pigmentosa 42.
Specifications
| KIAA1429 | |
| Polyclonal | |
| Unconjugated | |
| VIRMA | |
| 1110037F02Rik; 4930422M05Rik; DKFZP434I116; fSAP121; functional spliceosome-associated protein 121; KIAA1429; mKIAA1429; MSTP054; protein virilizer homolog; RGD1559904; RIKEN cDNA 1110037F02 gene; similar to mKIAA1429 protein; Virma | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 25962 | |
| -20°C | |
| Lyophilized |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 500 μg/mL | |
| PBS with 4mg trehalose and no preservative | |
| Q69YN4 | |
| VIRMA | |
| E.coli-derived human KIAA1429/VIRMA recombinant protein (Position: F11-K1529). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.