missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ KIAA1468 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15706911 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15706911 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15706911 Supplier Invitrogen™ Supplier No. PA559039

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: EAYSALSCCF ISEDLMVNHF LPGLRCLRTD MEHLSPEHEV ILSSMIKECE QKVENKTVQE PQGSMSIAAS LVSEDTKTKF Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen KIAA1468
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene KIAA1468
Gene Accession No. Q9P260
Gene Alias 2310035C23Rik; 6430401N10; HsT3308; HsT885; Kiaa1468; lisH domain and HEAT repeat-containing protein KIAA1468; lisH domain and HEAT repeat-containing protein KIAA1468 homolog; mKIAA1468; RAB11 binding and LisH domain, coiled-coil and HEAT repeat-containing; RAB11-binding protein containing LisH, coiled-coil, and HEAT repeats; RAB11-binding protein RELCH; Relch; RGD1307235; RIKEN cDNA 2310035C23 gene; similar to RIKEN cDNA 2310035C23
Gene Symbols KIAA1468
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human KIAA1468. Recombinant protein control fragment (Product #RP-98302).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57614
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.