missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Immunogen sequence: PVLENVSYMK QPAGASPSTL PSHVPGHAKL EKEQAAALGP ASATPALSSS PPPPSTLYRT QSPHGYPKSH STS Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 75%.
Activates PI3K and concomitantly recruits the WAVE1 complex to the close vicinity of PI3K and regulates neuronal morphogenesis.
Specifications
Specifications
| Antigen | KIAA1486 |
| Applications | Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | NYAP2 |
| Gene Accession No. | Q9P242 |
| Gene Alias | 9430031J16Rik; Jr6; Kiaa1486; neuronal tyrosine phosphorylated adaptor for phosphoinositide 3-kinase adaptor 2; neuronal tyrosine-phophorylated phosphoinositide 3-kinase adaptor 2; neuronal tyrosine-phosphorylated phosphoinositide-3-kinase adapter 2; neuronal tyrosine-phosphorylated phosphoinositide-3-kinase adaptor 2; NEWGENE_1305560; NYAP2 |
| Gene Symbols | NYAP2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?