missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Rat Troponin T Type 3 (fast skeletal) ELISA Kit (Colorimetric)
Shop All Bio Techne ProductsDescription
The Rat Troponin T Type 3 (fast skeletal) ELISA Kit (Colorimetric) measures rat Troponin T Type 3 (fast skeletal) in serum, plasma and other biological fluids.
Specifications
Specifications
| Assay Range | 31.25 - 2000 pg/mL |
| Assay Sensitivity | 18.75 pg/mL |
| Product Type | ELISA Kit (Colorimetric) |
| Sample Type | Biological Fluids, Plasma, Serum |
| Specificity | This kit recognizes Rat TNNT3 in samples. No significant cross-reactivity or interference between Rat TNNT3 and analogues was observed. |
| Gene ID (Entrez) | 7140 |
| Gene Symbol | TNNT3 |
| Host Species | Rat |
| Quantity | 1 Kit |
| Storage Requirements | Storage of components varies. See protocol for specific instructions. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?