missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIAA1522 Polyclonal Antibody

KIAA1522 Polyclonal Antibody
Brand: Invitrogen PA565117
This item is not returnable.
View return policy
Description
KIAA1522 Polyclonal Antibody for Western Blot, ICC/IF, IHC
Specifications
| KIAA1522 | |
| Polyclonal | |
| Unconjugated | |
| KIAA1522 | |
| KIAA1522; uncharacterized protein KIAA1522 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 57648 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9P206 | |
| KIAA1522 | |
| Recombinant Human KIAA1522. Recombinant protein control fragment (Product #RP-105879). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction