missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIAA1524 Monoclonal Antibody (OTI9E6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA808455S
This item is not returnable.
View return policy
Description
KIAA1524 Monoclonal antibody specifically detects KIAA1524 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| KIAA1524 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q8TCG1 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 57650 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI9E6 | |
| Unconjugated | |
| Cip2a | |
| AA408511; AU018569; C330027C09Rik; Cancerous inhibitor of PP2A; cancerous inhibitor of protein phosphatase 2A; Cip2a; Kiaa1524; p90; p90 autoantigen; p90 autoantigen homolog; Protein CIP2A; RGD1310335; RIKEN cDNA C330027C09 gene; similar to RIKEN cDNA C330027C09 | |
| Human recombinant protein fragment corresponding to amino acids 635-905 of human KIAA1524 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction