missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KIAA1549 Polyclonal Antibody

Description
This target displays homology in the following species: Cow: 93%; Dog: 93%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 93%; Rabbit: 93%; Rat: 93%.
KIAA1549 is a protein coding gene.
Specifications
Specifications
| Antigen | KIAA1549 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | KIAA1549 |
| Gene Accession No. | Q68FD9, Q9HCM3 |
| Gene Alias | D630045J12Rik; KIAA1549; KIAA1549 ortholog; mKIAA1549; RIKEN cDNA D630045J12 gene; UPF0606 protein KIAA1549; UPF0606 protein KIAA1549 homolog; UPF0606 protein KIAA1549; UPF0606 protein KIAA1549 homolog |
| Gene Symbols | D630045J12Rik, KIAA1549 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?