Learn More
Invitrogen™ KIAA1609 Monoclonal Antibody (OTI12B1)
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA525070
Product Code. 15954674
  • € 588.00 / 100µL
Description
KIAA1609 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
TLDC1 (TBC/LysM-Associated Domain Containing 1) is a Protein Coding gene. An important paralog of this gene is NCOA7.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
KIAA1609
Monoclonal
1.0 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q6P9B6
TLDC1
Full length human recombinant protein of KIAA1609 produced in HEK293T cell.
100 μL
Primary
Human
Antibody
IgG1
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
OTI12B1
Unconjugated
TLDC1
4632415K11Rik; Kiaa1609; MEAK7; MTOR associated protein, eak-7 homolog; MTOR-associated protein MEAK7; TBC/LysM associated domain containing 1; TBC/LysM-associated domain containing 1; TBC/LysM-associated domain-containing protein 1; TLD domain containing 1; TLD domain-containing protein 1; TLDC1
Mouse
Affinity Chromatography
RUO
57707
-20°C, Avoid Freeze/Thaw Cycles