missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIAA1958 Antibody [Janelia Fluor™ 585], Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-20225JF585
Denne vare kan ikke returneres.
Se returpolicy
Beskrivelse
KIAA1958 Polyclonal antibody specifically detects KIAA1958 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Tekniske data
| KIAA1958 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 158405 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| Janelia Fluor 585 | |
| FLJ39294, hypothetical protein LOC158405, KIAA1958, MGC142075 | |
| Produced in rabbits immunized with E. coli-derived Human KIAA1958 fragment | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion