missing translation for 'onlineSavingsMsg'
Learn More

CoraLite™ Plus 488 Anti-Human CD5 (L17F12), Proteintech

Product Code. 30236806 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
100 Tests
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30236806 100 Tests 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30236806 Supplier Proteintech Supplier No. CL48865200100TESTS

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

The CD5 antibody from Proteintech is a mouse monoclonal antibody generated with human t-cell acute lymphoblastic leukemia (t-all) cells of human CD5. This antibody recognizes human antigen. The CD5 antibody has been validated for the following applications: FC analysis. This antibody is coralite™ plus 488.

Specifications

Antigen CD5
Applications Flow Cytometry
Classification Monoclonal
Clone L17F12
Concentration 5 μl/test
Conjugate CoraLite Plus 488
Gene Symbols LEU1; T1
Host Species Mouse
Immunogen Human T-cell acute lymphoblastic leukemia (T-ALL) cells
Purification Method Protein A purification
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 921
Target Species Human
Content And Storage Store at 2-8°C. Avoid exposure to light. Stable for one year after shipment.
Product Type Flow Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.