missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIBRA Antibody (OTI1H8), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71732B
This item is not returnable.
View return policy
Description
KIBRA Monoclonal specifically detects KIBRA in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| KIBRA | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| WWC1 | |
| Full length recombinant protein of human KIBRA (NP_056053) produced in HEK293T cells. | |
| 0.1 mL | |
| 23286.0 | |
| Store at 4°C in the dark. |
| Western Blot | |
| OTI1H8 | |
| Western Blot | |
| FLJ10865, FLJ23369, HBeAg-binding protein 3, KIAA0869HBEBP3, KIBRAHBEBP36, Kidney and brain protein, protein KIBRA, protein WWC1, WW and C2 domain containing 1, WW domain-containing protein 1, WW, C2 and coiled-coil domain containing 1 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction