missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KID Polyclonal Antibody

Description
Immunogen sequence: LEPEEKAEDCW ELQISPELLA HGRQKILDLL NEGSARDLRS LQRIGPKKAQ LIVGWRELHG PFSQVEDLER VEGITGKQME SF Highest antigen sequence identity to the following orthologs - mouse 83%, rat 81%.
The protein encoded by this gene is a member of kinesin-like protein family. This family of proteins are microtubule-dependent molecular motors that transport organelles within cells and move chromosomes during cell division. The C-terminal half of this protein has been shown to bind DNA. Studies with the Xenopus homolog suggests its essential role in metaphase chromosome alignment and maintenance.
Specifications
Specifications
| Antigen | KID |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.4 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | KIF22 |
| Gene Accession No. | Q14807 |
| Gene Alias | A-328A3.2; AU021460; C81217; Kid; Kif22; Kif22a; kinesin family member 22; kinesin family member 22A; kinesin-like 4; Kinesin-like DNA-binding protein; kinesin-like DNA-binding protein pseudogene; kinesin-like protein 4; Kinesin-like protein KIF22; KNSL4; OBP; OBP-1; OBP-2; origin of plasmid DNA replication-binding protein; oriP binding protein; SEMDJL2 |
| Gene Symbols | KIF22 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?