missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIDINS220 Antibody, Alexa Fluor™ 700, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-50035AF700
This item is not returnable.
View return policy
Description
KIDINS220 Polyclonal antibody specifically detects KIDINS220 in Human samples. It is validated for Western Blot
Specifications
| KIDINS220 | |
| Polyclonal | |
| Western Blot | |
| kinase D-interacting substrate, 220kDa | |
| The immunogen for this product maps to a region between residue 1721 and 1771 of human Ankyrin Repeat-Rich Membrane Spanning using the numbering given in entry NP_065789.1 (GeneID 57498). | |
| 0.1 mL | |
| Phospho Specific | |
| 57498 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction