Learn More
KIDINS220 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-50035
This item is not returnable.
View return policy
Description
KIDINS220 Polyclonal specifically detects KIDINS220 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| KIDINS220 | |
| Polyclonal | |
| Unconjugated | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
| KIDINS220 | |
| The immunogen for this product maps to a region between residue 1721 and 1771 of human Ankyrin Repeat-Rich Membrane Spanning using the numbering given in entry NP_065789.1 (GeneID 57498). | |
| 100 μL | |
| Primary | |
| Human | |
| IgG |
| Western Blot, Immunoprecipitation | |
| 1.0 mg/mL | |
| Western Blot 1:100-1:2000, Immunoprecipitation 2-5 mcg/mg | |
| kinase D-interacting substrate, 220kDa | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 57498 | |
| Store at 4C. Do not freeze. |