missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIF1-binding protein Antibody, FITC, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97110F
This item is not returnable.
View return policy
Description
KIF1-binding protein Polyclonal specifically detects KIF1-binding protein in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| KIF1-binding protein | |
| Polyclonal | |
| ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| DKFZP586B0923, KBP, KIAA1279, KIF1-binding protein, TTC20 | |
| Produced in rabbits immunized with purified, recombinant KIF1-binding protein (Q96EK5; Met1-Thr621). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| FITC | |
| PBS | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 26128 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction