missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ KIF11 Recombinant Rabbit Monoclonal Antibody (6G8V8)
GREENER_CHOICE

Product Code. 17136213
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17136213 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17136213 Supplier Invitrogen™ Supplier No. MA535554

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: ETIKIGLTKL NCFLEQDLKL DIPTGTTPQR KSYLYPSTLV RTEPREHLLD QLKRKQPELL MMLNCSENNK EETIPDVDVE EAVLGQYTEE PLSQEPSVDA GVDCSSIGGV PFFQHKKSHG KDKENRGINT LERSKVEETT EHLVTKSRLP LRAQINL.

This gene encodes a motor protein that belongs to the kinesin-like protein family. Members of this protein family are known to be involved in various kinds of spindle dynamics. The function of this gene product includes chromosome positioning, centrosome separation and establishing a bipolar spindle during cell mitosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KIF11
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 6G8V8
Concentration 0.53 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene KIF11
Gene Accession No. P52732
Gene Alias Eg5; HKSP; KIF11; Kif8; Kifl1; Kines; kinesin 11; kinesin family member 11; kinesin-like 1; kinesin-like protein 1; Kinesin-like protein KIF11; kinesin-like spindle protein HKSP; Kinesin-related motor protein Eg5; Knsl1; MCLMR; Thyroid receptor-interacting protein 5; TR-interacting protein 5; TRIP5; TRIP-5
Gene Symbols KIF11
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 900-1056 of human EG5/KIF11 (P52732).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3832
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.