missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ KIF13B Polyclonal Antibody
GREENER_CHOICE

Product Code. 16360925
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16360925 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16360925 Supplier Invitrogen™ Supplier No. PA583973

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: LPSGKNDGSI GGKQYFRCNP GYGLLVRPSR VRRATGPVRR RSTGLRLGAP EARRSATLSG SATNLASLTA ALAKADRSHK NPENRKSWAS Highest antigen sequence indentity to the following orthologs: Mouse - 87%, Rat - 87%.

KIF13B (Kinesin Family Member 13B) is a Protein Coding gene. Diseases associated with KIF13B includeSpastic Paraplegia 37, Autosomal DominantandSpastic Paraplegia 11, Autosomal Recessive. Among its related pathways areGolgi-to-ER retrograde transportandFactors involved in megakaryocyte development and platelet production. Gene Ontology (GO) annotations related to this gene includeprotein kinase bindingandmicrotubule motor activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KIF13B
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene KIF13B
Gene Accession No. Q9NQT8
Gene Alias 5330429L19Rik; 6030414C01; AI429803; C130021D12Rik; GAKIN; guanylate kinase associated kinesin; KIAA0639; Kif13b; kinesin 13B; kinesin family member 13B; kinesin-like protein GAKIN; Kinesin-like protein KIF13B; N-3 kinesin
Gene Symbols KIF13B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human KIF13B. Recombinant protein control fragment (Product #RP-108203).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23303
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.