missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIF1B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
Brand: Novus Biologicals NB100-57493
This item is not returnable.
View return policy
Description
KIF1B Polyclonal specifically detects KIF1B in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| KIF1B | |
| Polyclonal | |
| Unconjugated | |
| TBS and 0.1% BSA with 0.09% Sodium Azide | |
| CMT2, CMT2A, CMT2A1, HMSNII, KIAA0591FLJ23699, KIAA1448, kinesin family member 1B, kinesin superfamily protein KIF1B, kinesin-like protein KIF1B, Klp, MGC134844, NBLST1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| IgG |
| Western Blot, Immunoprecipitation | |
| 0.2 mg/mL | |
| Western Blot 1:2000-1:10000, Immunoprecipitation 2-5 ug/mg lysate | |
| O60333 | |
| KIF1B | |
| The immunogen recognized by this antibody maps to a region between residue 1575 and 1625 of human kinesin family member 1B using the numbering given in entry CAC16629.1 (GeneID 23095). | |
| 0.1 mL | |
| Stem Cell Markers | |
| 23095 | |
| Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction