missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KIF1B Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA576971
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
The kinesins constitute a large family of microtubule-dependent motor proteins, which are responsible for the distribution of numerous organelles, vesicles and macromolecular complexes throughout the cell. Individual kinesin members play crucial roles in cell division, intracellular transport and membrane trafficking events including endocytosis and transcytosis. KIF1B is a member of the KIF1/Unc104 family of kinesin-like proteins that are involved in the transport of mitochondria or synaptic vesicles in axons. KIF1B is an amino-terminal-type motor protein that is ubiquitously expressed, with the most abundant levels in differentiated nerve cells. The human KIF1B gene maps to chromosome 1p36. 2. Defects in axonal transport due to mutations at the KIF1B gene can underlie the human peripheral neuropathy phenotype. The mouse KIF1B gene generates an alternatively spliced transcript, which produces two isoforms.
Specifications
| KIF1B | |
| Polyclonal | |
| Unconjugated | |
| KIF1B | |
| A530096N05Rik; AI448212; AI506502; CMT2; CMT2A; CMT2A1; D4Mil1e; HMSNII; KIAA0591; KIAA1448; Kif1b; KIF1Bp130; KIF1Bp204; kinesin family member 1B; kinesin heavy chain member 1B; kinesin superfamily protein KIF1B; kinesin-family protein KIF1Bbeta3; kinesin-like protein KIF1B; Klp; N-3 kinesin; NBLST1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 117548, 23095 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| TBS with 1% BSA, 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| O60333, O88658 | |
| KIF1B | |
| KLH conjugated synthetic peptide derived from human KIF1B: 13-120. | |
| 100 μL | |
| Primary | |
| Human, Rat, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction