missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIF25 Antibody (OTI2F4), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71077AF350
This item is not returnable.
View return policy
Description
KIF25 Monoclonal specifically detects KIF25 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| KIF25 | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| KIF25 | |
| Full length recombinant protein of human KIF25 (NP_005346) produced in HEK293T cells. | |
| 0.1 mL | |
| 3834.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI2F4 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| kinesin family member 25 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction